Principaux documents
CLS3640
Corning® NBS 384 well microplates
surface treatment, Non-binding Surface, clear polystyrene, flat bottom, non-sterile, lid: no, pack of 25
Synonyme(s) :
384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.15
Matériaux
clear polystyrene
flat bottom
flat bottom wells clear wells
polystyrene
square clear wells
Stérilité
non-sterile
non-sterile
Caractéristiques
lid: no
lid: no
skirt
plate format: 384 well standard
Conditionnement
case of 100
pack of 25
Fabricant/nom de marque
Corning 3640
Volume maximum
112 μL
Taille
384 wells
Volume de travail
80 μL
Type de liaison
NBS (non-binding surface)
Description générale
  • Nonbinding (NBS) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions
  • Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise
Non-Binding surface (NBS) multiwell plates have a modified polymer surface (PS) which results in a non-ionic hydrophilic surface (polyethylene oxide-like), thereby minimizing bio-interaction and enhancing the signal-to-noise ratio. Results have demonstrated NBS′s ability to significantly reduce protein and nucleic acid binding to polymers (<2 ng/cm2; over 90% reduction of protein binding compare to PS & PP surface), maintain enzyme activity, and inhibit adhesion of a number of cell lines. These novel surface technologies for assay plates are ideal for homogeneous assays such as Scintillation Proximity Assay (SPA). In addition to a 20+ fold increase in signal-to-noise ratio, selected assays have also displayed improved sensitivity and specificity. The recommended working volume of up to 20 μL to 80 μL for 384 well plates.
Informations légales
Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.