Principaux documents
CLS11405
Corning® Erlenmeyer flask, with aseptic connector
volume 125 mL, I.D. 1/8 in., sterile
Synonyme(s) :
Glass Erlenmeyer flask
About This Item
Code UNSPSC :
41122104
Nomenclature NACRES :
NB.43
Matériaux
flat bottom flask
polycarbonate
polypropylene cap
Description
plain
Stérilité
sterile
Caractéristiques
cap: no
C-Flex tubing
Conditionnement
case of 6
Fabricant/nom de marque
Corning 11405
D.I.
1/8 in.
Capacité
125 mL
Volume
125 mL
Raccords
male luer lock
Caractéristiques et avantages
  • An aseptic transfer cap is preassembled on an optically clear polycarbonate flask
  • Graduated for better accuracy
  • Cap is made of flexible tubing that is heat-sealable and chemically resistant with male luer lok disconnect
  • Gamma-radiation sterilized
  • When adding or removing medium and cells, this flask enhances the productivity of the shaker culture operation
  • Nonpyrogenic
Autres remarques
Informations légales
Corning is a registered trademark of Corning, Inc.