ZLXLPLUGP
Outil de régénération de l'OI
plug, For water purification systems with two Progard® Packs installed
About This Item
Code UNSPSC :
41104200
Nomenclature NACRES :
JA.13
Produits recommandés
Conditionnement
pkg of 1 unit
Compatibilité
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
Description générale
The RO Regeneration Tool/Plug is used along with the RO Regeneration Tool (ZLXLCLPAK) when the system has 2 Progard® Packs installed. RO Regeneration tools are used in place of real Progard® packs whenever a RO cleaner (Cl2 tablets, RO Clean-A and RO Clean-B) is being used.
Application
Used for the pH cleaning of the system using chlorine tablets when 2 Progard® Packs are installed.
Caractéristiques et avantages
Genuine accessory for RO Regeneration Tool/Plug - ZLXLP0D01
Informations légales
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
PROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany