Principaux documents
Z693731
LabJaws jumbo clamp holder
stainless steel
Synonyme(s) :
clamp holders, frame connectors
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.35
Matériaux
stainless steel
Fabricant/nom de marque
Troemner 916357
Capacité de la mâchoire
0-18 mm
Description générale
Corrosion resistant. Has precise fit and powerful holding ability for heavy-duty applications. Ideal for holding clamps to lab frames or ring stands.
Informations légales
LabJaws is a trademark of Henry Troemner LLC