Principaux documents
Z680826
Duran® stirred reactor caps and accessories, for GL 80 stirred reactor systems
anchor type magnetic stirrer.
Synonyme(s) :
1200383
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Conditionnement
pack of 1 ea
Fabricant/nom de marque
DURAN® 1200383
Diamètre
62 mm
Description générale
These connections fit the GL 80 stirred reactor systems which provide addition or removal of media from the bottle during the mixing process.
  • The accessories can be autoclaved and are suitable for use in the biological sector
  • The agitating elements are exchangeable and use up to 500 rpm is possible
Caractéristiques et avantages
• Manufactured from chemically resistant materials: PP, PTFE, PEEK, stainless steel• Drive with standard magnetic stirrers up to 500 rpm and allows improved mixing compared to stir bars• The variable stirrer shaft can be vertically adjusted to the optimal position in either 500, 1000 or 2000 ml DURAN® GLS 80® bottles• A shorter stirrer shaft is available for the 250 ml DURAN® GLS 80® bottles• The 4x threaded ports (2x GL 14 and 2x GL 18) can be blanked off or used for connecting tubing or measurement probes• Flexible: The proven DURAN® GL 45 / GLS 80 connection system components (2x GL 14 and 2x GL 18) can be used with 1.6 to 12.0 mm outer diameter semi-rigid tubing to add or remove liquid or gas during the mixing process• Fully Autoclavable components
Informations légales
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group