Z674540
Duran® GLS 80 caps and accessories
Insert for GL 18 screw cap, I.D. 12.0 mm
Synonyme(s) :
1160173
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Produits recommandés
Fabricant/nom de marque
DURAN® 1160173
Description générale
Flexible modular liquid or gas transfer 80 connection cap system. Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation is possible by using the syringe membrane filter. Unused ports can be closed with a closed top GL 18 PBT cap.
Informations légales
DURAN is a registered trademark of DWK Life Sciences