Principaux documents
Z566284
Clamping rings for glove box ports
port O.D. 6 in.
Synonyme(s) :
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
About This Item
Code UNSPSC :
42132201
Nomenclature NACRES :
NB.35
Caractéristiques
for glove boxes and Techni-Dome® 6 in. O.D.
Conditionnement
pack of 2 ea
Fabricant/nom de marque
Bel-Art H50029-0200
Description générale
Clamping rings fit 6 in. O.D. glove ports for the Techni-Dome® glove chamber, Z564869.
Informations légales
Techni-Dome is a registered trademark of SP Scienceware, a division of SP