Principaux documents
Z565016
Replacement parts for Buchi® vacuum controllers and pumps
Control cable between vacuum controller V850/855 and vacuum pump V700/710
About This Item
Code UNSPSC :
41105101
Nomenclature NACRES :
NB.43
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Büchi 44989
Longueur
2 m
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG