Principaux documents
Z563099
J-KEM® thermocouple probe
Single-element, thermocouple type, T, diam. × L 1/8 in. × 6 in.
About This Item
Code UNSPSC :
41112303
Nomenclature NACRES :
NB.76
Description
PTFE-coated
Caractéristiques
thermocouple type T
Fabricant/nom de marque
J-Kem
Diam. × L
1/8 in. × 6 in.
Informations légales
J-KEM is a registered trademark of J-KEM Electronics, Inc.