Principaux documents
Z516201
Sigma-Aldrich
Aldrich® short-path distillation head with jacketed condenser
Joints: ST/NS 29/32 (2), thermometer joint: ST/NS 10/30
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Joint
thermometer joint: ST/NS10/30
Joints: ST/NS29/32 (2)
Condenseur, L
300 mm
Profondeur d'immersion
75 mm
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC