Z421960
Aldrich® three-neck round-bottom flask
vertical, capacity 12,000 mL, center joint: ST/NS 34/45, side Joints: ST/NS 24/40 (2)
Synonyme(s) :
flask round bottom, round bottom flasks, round bottomed flask
About This Item
Produits recommandés
Matériaux
glass
round bottom flask
three-neck (Vertical)
Joint
center joint: ST/NS34/45
side Joints: ST/NS24/40 (2)
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Sigma-Aldrich
Capacité
12,000 mL