Z302368
Quickfit® Cone/Screw-thread Adapters
13 thread (couples 6.0 - 7.0 mm diameter), joint: ST/NS 29/32
Synonyme(s) :
ST54/13
About This Item
Produits recommandés
Joint
joint: ST/NS29/32
Fabricant/nom de marque
Quickfit ST 54/13
Disponibilité
not available in USA and Canada
not available in USA and Canada
Raccords
13 thread (couples 6.0 - 7.0 mm diameter)
Description générale
Can be used in apparatus at temperatures up to 300°C providing excessive heat is not applied directly to the screw-thread union.
For replacement parts, see related products section of web site listing.