Principaux documents
Z286478
Nalgene® stopcock
3-way, bore size 4 mm
Synonyme(s) :
3-way connector, hose connector
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.79
Matériaux
polypropylene (with TFE plug)
Description
for tubing I.D. 1/4 -5/16 in.
Caractéristiques
autoclavable
Conditionnement
pack of 1 ea
case of 4 ea
Fabricant/nom de marque
Nalgene® 6470, 0004
Taille de l'alésage
4 mm
Description générale
Serrated tubulation on each end.
Caractéristiques et avantages
  • Autoclave friendly
  • Non-seizing, leakproof PTFE plug
  • Tubulated outlets are arranged in a T shape and accepts tubing of 1/4 in. 7/16 in.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries