Principaux documents
Z280356
Sigma-Aldrich
PVC laboratory tubing
I.D. × O.D. 1/8 in. × 1/4 in.
Synonyme(s) :
Saint Gobain Versilon ABH02007
About This Item
Code UNSPSC :
41105108
Nomenclature NACRES :
NB.35
Conditionnement
pkg of 100 feet
D.I. × D.E.
1/8 in. × 1/4 in.
Taille de la paroi
1/16 in.
Description générale
General-purpose polyvinyl chloride laboratory tubing. Clear, flexible tubing ideal for drains and rinse tubes. Max. temp.: 165°F (73.4°C).