Principaux documents
Z219800
Sigma-Aldrich
Single-bulb ball-tube
capacity 250 mL, joint: ST/NS 24/40, w/drip-tip
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS24/40
Capacité
250 mL
Description générale
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.