Principaux documents
Z219789
Sigma-Aldrich
Single-bulb ball-tube
capacity 250 mL, joint: ST/NS 14/20, w/drip-tip
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS14/20
Capacité
250 mL
Catégories apparentées
Description générale
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.