Principaux documents
Z190543
Sigma-Aldrich
Liquid scintillation vials (with screw cap attached)
urethane foam-filled high-density polyethylene cap, liner (Poly-Seal cone), pkg of 500 ea
Synonyme(s) :
Scintillation Counting Vials
About This Item
Code UNSPSC :
41121806
Nomenclature NACRES :
NB.11
Matériaux
clear glass vial
liner (Poly-Seal cone)
urethane foam-filled high-density polyethylene cap
Caractéristiques
closure type screw top vial
closure type white polyethylene cap
Conditionnement
pkg of 500 ea
Taille
28 mm × 61 mm
Volume
20 mL
Raccords
thread for 22-400
Description générale
Autoclavable, easy to maintain. A vial is used to store or as a closure type,in order to prevent contamination.
Reliable Wheaton "180" brand low-potassium glass. White 22 mm caps have tops suitable for writing. Vials with caps attached are packed in individual cells in trays of 100.