Principaux documents
Z127779
Sigma-Aldrich
Aldrich® fraction cutter head
joint: ST/NS 24/40
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS24/40
Description générale
50 mL barrel, with 4 mm bore glass stopcocks.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC