Principaux documents
Z116645
Sigma-Aldrich
Aldrich® gas-condensing bottle
with Luer lock
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Matériaux
cap (Luer lock)
Conditionnement
pack of 1
Fabricant/nom de marque
Sigma-Aldrich
Capacité
150 mL
Description générale
With stopcock, graduated, holds 150 mL of liquefied gas.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC