Principaux documents
THS01115
ThalesNano CatCart® catalyst cartridge system, 30 mm L
20%Pd(OH)2/C
Synonyme(s) :
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
Code UNSPSC :
60104500
Nomenclature NACRES :
NB.51
Conditionnement
pack of 6 ea
Fabricant/nom de marque
ThalesNano THS 01115
Diamètre
4 mm
Description générale
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Informations légales
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.