Principaux documents
SYNM211042
Synthware nitrogen/argon line manifold with PTFE stopcocks
port size 4, Hose Connections: left-right, stopcock size 2 mm
Synonyme(s) :
Synthware M211042, vacuum manifold, vacuum unit
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware M211042
Taille du port
4
Taille du robinet d'arrêt
2 mm
Description générale
With 14/20 outer joint at the center to fit a rubber septum for keeping syringes under an inert environment cutting down on the number used. Main tube is 22 mm O.D. with both ends connections(10 mm). Delivery ports have 8 mm hose connections.
Informations légales
Synthware is a trademark of Kemtech America Inc.