Principaux documents
SYNF564250
Synthware reaction tube with high vacuum valve
250 mL, joint: ST/NS 24/40, valve size 0-4 mm
Synonyme(s) :
Synthware F564250, Schlenk flask
About This Item
Code UNSPSC :
41121700
Nomenclature NACRES :
NB.51
Matériaux
colorless tube
glass tube
Joint
joint: ST/NS24/40
Stérilité
non-sterile
Caractéristiques
graduations: no
Push fit
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Kemtech F564250
Capacité
250 mL
250 mL
Taille de la vanne
0-4 mm
Description générale
Supplied with high vacuum valve with PTFE o-ring at tip and PTFE wiper (“umbrella”) to protect Viton® O-ring on shaft.
Informations légales
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company