Principaux documents
SYNF459125
Synthware Squibb-style separatory funnel with standard taper joints, PTFE stopcock and PTFE stopper
125 mL, joint: ST/NS 19/22, PTFE stopcock and PTFE stopper
Synonyme(s) :
Synthware F459125, glass separatory funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F459125
Capacité
125 mL
125 mL
Description générale
With 2 mm PTFE stopcock. Supplied with a glass stopper or PTFE stopper.
Informations légales
Synthware is a trademark of Kemtech America Inc.