Principaux documents
SYNF164100
Synthware two neck reaction flask with glass stopcock
capacity 100 mL, joint: ST/NS 24/40
Synonyme(s) :
Synthware F164100, Glass reaction flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
round bottom
two-neck
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F164100
Capacité
100 mL
Description générale
With a 2 mm glass stopcock having a constant pressure retention device.
Informations légales
Synthware is a trademark of Kemtech America Inc.