Principaux documents
SYNA381420
Synthware 90 degree vacuum adapter with PTFE stopcock
male joint: ST/NS 14/20
Synonyme(s) :
Synthware A381420, glass adapter with stopcock
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
male joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A381420
Description générale
With 8 mm or 10 mm O.D. hose connection and 2 mm PTFE stopcock.
Informations légales
Synthware is a trademark of Kemtech America Inc.