Principaux documents
BR780760
BRAND® micro tubes without tamper-evident screw cap
capacity 0.5 mL, self-standing bottom, non-sterile
Synonyme(s) :
Micro Tubes with Screw caps, Microcentrifuge Tubes, Tubes
About This Item
Code UNSPSC :
41121702
Nomenclature NACRES :
NB.43
Matériaux
colorless tube
polypropylene tube
self-standing bottom
Stérilité
non-sterile
Conditionnement
pack of 1000 ea
Fabricant/nom de marque
BRAND 780760
Capacité
0.5 mL
Diam. du tube
13 mm
Description générale
Operating range: -196 °C to +121 °C. Autoclavable at 121 °C (2 bar), acc. DIN EN 285.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG