Principaux documents
BR472000
BRAND® staining tray for glass staining trough
soda-lime glass
Synonyme(s) :
Microscopy, Staining, Staining Accessories
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
MA.01
Matériaux
soda-lime glass
Description
use with glass staining trough
Fabricant/nom de marque
BRAND 472000
L × l × H
88 mm × 70 mm × 40 mm, holds 10 slides
Description générale
staining trough, tray, and wire handle are sold separately
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG