Merck
Exploring the radiosynthesis and in vitro characteristics of [
Journal of labelled compounds & radiopharmaceuticals (2017-05-31)
Svend B Jensen, Meeri Käkelä, Lars Jødal, Olli Moisio, Aage K O Alstrup, Sirpa Jalkanen, Anne Roivainen
ABSTRACT
Vascular adhesion protein 1 is a leukocyte homing-associated glycoprotein, which upon inflammation rapidly translocates from intracellular sources to the endothelial cell surface. It has been discovered that the cyclic peptide residues 283-297 of sialic acid-binding IgG-like lectin 9 (Siglec-9) "CARLSLSWRGLTLCPSK" bind to vascular adhesion protein 1 and hence makes the radioactive analogues of this compound ([
Sigma-Aldrich
Porcine Serum, USA origin, sterile-filtered, suitable for cell culture