Merck
  • Analysis of N-acetylaminosugars by CE: a comparative derivatization study.
Analysis of N-acetylaminosugars by CE: a comparative derivatization study.
Electrophoresis (2009-07-22)
Isabella Rustighi, Cristiana Campa, Marco Rossi, Sabrina Semeraro, Amedeo Vetere, Amelia Gamini
ABSTRACT
N-linked or O-linked glycans derived from glycoprotein processing carry, an N-acetylglucosamine or an N-acetylgalactosamine respectively, at their reducing termini. The presence of the N-acetylamino group on C-2 of reducing sugar residues has been reported to hamper the derivatization reaction with a chromophore at the anomeric centre. In this paper N-acetyllactosamine, N-acetylglucosamine, N-acetylgalactosamine and several other neutral monosaccharides are coupled to three different dyes (4-aminobenzonitrile, 2-aminopyridine, 2-aminobenzoic acid (2-AA)) by reductive amination and analysed by CE with UV detection. The 2-AA derivatives showed the lowest concentration detection limits, varying approximately in the 2-3 muM range for the saccharides tested including the N-acetamido ones. The possibility to separate and detect with the same sensitivity ten 2-AA-labelled monosaccharides mainly found in mammalian or plant glycoproteins in a single CE run is highlighted. The analysis has been carried out in less than 25 min using the borate-complexation method in CZE mode. The influence of the strength of the acid used as catalyst in the chemical modification of the sugars with 2-AA is also shortly addressed.
Sigma-Aldrich
4-Aminobenzonitrile, 98%