24445
SGE Needles for RN syringes
volume 10 μL, N10-VA8X00H-II, needle size 25 ga, needle L × O.D. 53 mm × 0.50 mm
Synonym(s):
REPL.NEEDLE N10-VA8100H,EA
About This Item
Recommended Products
material
cone tip, side port stainless steel needle (point style 5)
needle size
25 ga
feature
needle cone tip, side-port
packaging
pkg of 1 ea
manufacturer/tradename
SGE 037777
needle L × O.D.
53 mm × 0.50 mm
volume
10 μL
compatibility
for use with 10RVA8X, 8100, and 8200
Looking for similar products? Visit Product Comparison Guide
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service