Skip to Content
Merck
All Photos(1)

Key Documents

SML2554

Sigma-Aldrich

RIPK2 inhibitor 1

≥98% (HPLC)

Synonym(s):

3-(Benzoylamino)-4-methyl-N-[3-(1-methyl-1H-imidazol-2-yl)phenyl]-benzamide, MolPort-016-359-762

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22N4O2
CAS Number:
Molecular Weight:
410.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to very dark gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC=C(C(NC2=CC(C3=NC=CN3C)=CC=C2)=O)C=C1NC(C4=CC=CC=C4)=O

Biochem/physiol Actions

RIPK2 inhibitor 1 is a potent and selective inhibitor of Receptor interacting protein kinase 2 (RIPK2) that potently inhibit NFκB activity and the proliferation of cancer cell lines. RIPK2 inhibitor 1 inhibits intestinal and lung inflammation in rodent models.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service