Skip to Content
Merck
All Photos(1)

Key Documents

PZ0144

Sigma-Aldrich

PD-161989 2-Hydroxyethanesulfonate

≥98% (HPLC)

Synonym(s):

1,4-Dihydro-6-methyl-7-nitro-5-(1-pyrrolodinemethyl)-2,3-quinoxaline 2-hydroxyethanesulfonic acid salt, PD-161989 2-hydroxyethanesulfonic acid salt, PD-161989 isethionate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N4O4 · C2H6O4S
CAS Number:
Molecular Weight:
430.43
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

H2O: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

OCCS(O)(=O)=O.Cc1c(CN2CCCC2)c3NC(=O)C(=O)Nc3cc1[N+]([O-])=O

InChI

1S/C14H16N4O4.C2H6O4S/c1-8-9(7-17-4-2-3-5-17)12-10(6-11(8)18(21)22)15-13(19)14(20)16-12;3-1-2-7(4,5)6/h6H,2-5,7H2,1H3,(H,15,19)(H,16,20);3H,1-2H2,(H,4,5,6)

InChI key

KFHUCRVOUVNJSQ-UHFFFAOYSA-N

Biochem/physiol Actions

PD-161989 is an AMPA receptor antagonist.

Features and Benefits

This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service