Skip to Content
Merck
All Photos(1)

Key Documents

CLS3573

Corning® 384 well microplate, low flange

black polystyrene, flat bottom, non-sterile, lid: no, pack of 50

Synonym(s):

384 multiwell plates, 384 well immunoassay plates, 384 well microplates, 384 well microtiter plates, 384 well plates, immunoassay plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
black bottom
black polystyrene
flat bottom
polystyrene
square black wells
maximum volume:
112 μL
size:
384 wells
working volume:
80 μL
Pricing and availability is not currently available.

material

black bottom
black polystyrene
flat bottom
polystyrene
square black wells

sterility

non-sterile

feature

lid: no
skirt

packaging

pack of 50

manufacturer/tradename

Corning 3573

maximum volume

112 μL

size

384 wells

working volume

80 μL

color

black

suitability

suitable for (cell cuture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Black wall plates have reduced autofluorescence, higher signal to background ratio and lower CV′s.
  • Enhanced flatness provdes low intra- and inter- plate CV′s.
  • Conforms to standard microplate footprint and dimensions
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm2
  • Ideal for today′s more sensitive assays and plate readers

Note: Suggested replacement for CLS3708 and CLS3710

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service