Skip to Content
Merck
All Photos(1)

Key Documents

MABS2239

Sigma-Aldrich

Anti-CXCL9 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Pricing and availability is not currently available.

biological source

hamster

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2A6.9.9, monoclonal

mol wt

calculated mol wt 14.44 kDa
observed mol wt ~14 kDa

species reactivity

mouse

packaging

antibody small pack of 100 μL

technique(s)

neutralization: suitable
western blot: suitable

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
04243PX156592214
solubility

208 g/L

solubility

H2O: soluble 4.5 parts, alcohol: insoluble

solubility

soluble 208 g/L

solubility

-

assay

≥99.5% dry basis (alkalimetric), 99.5-100.5% (alkalimetric)

assay

98-100.5% (calc. to the dried substance)

assay

≥99% (acidimetric)

assay

-

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

100, 300

mp

252.6 °C (lit.)

mp

252.6 °C (lit.)

mp

252.6 °C (lit.)

mp

252.6 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

General description

C-X-C motif chemokine 9 (UniProt: P18340; also known as Gamma-interferon-induced monokine, Monokine induced by interferon-gamma, MIG, MuMIG, Protein m119, Small-inducible cytokine B9) is encoded by the Cxcl9 (also known as Scyb9) gene (Gene ID: 17329) in murine species. CXCL9 is a small cytokine of the CXC chemokine family that lacks the Glu-Leu-Arg (ELR) domain. It is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off to generate the mature form. It is induced in macrophages in response to interferon- g and participates in immune and inflammatory response. Its actions are mediated via CXC-chemokine receptor 3 (CXCR3). CXCL10 and CXCL11 that are structurally related to CXCL9 also serve as ligands for CXCR3 and contribute to its activation in CXCR3 expressing cells. Overexpression of CXCL9 is reported to reduced tumor progression and metastasis via the inhibition of angiogenesis. Conversely, CXCL9 can also act directly on tumor cells expressing the CXCR3 receptor to promote cell migration and epithelial mesenchymal transition.

Specificity

Clone 2A6 is an Armenian Hamster monoclonal antibody that detects murine CXCL9/MIG.

Immunogen

Recombinant mouse C-X-C motif chemokine 9 (CXCL9).

Application

Quality Control Testing

Evaluated by Western Blotting with murine recombinant CXCL9/MIG protein.

Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected recombinant murine CXCL9/MIG protein.

Tested Applications


Neutralizing Analysis: A representative lot detected CXCL9 in Neutralizing applications (Christen, U., et. al. (2003). J Immunol. 171(12):6838-45; Tajfirouz, D., et. al. (2017). Virology. 506:7-13).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CXCL9, clone 2A6.9.9, Cat. No. MABS2239, is an Armenian hamster monoclonal antibody that detects C-X-C motif chemokine 9 (CXCL9) and is tested for use in Neutralizing and Western Blotting.

Physical form

Purified Armenian hamster monoclonal antibody IgG in PBS without preservatives.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service