Skip to Content
Merck
All Photos(1)

Key Documents

8.18619

Sigma-Aldrich

ß-Ionone

for synthesis

Synonym(s):

ß-Ionone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-224-3
NACRES:
NA.22

vapor pressure

<1 hPa ( 25 °C)

Quality Level

Assay

≥95.0% (GC)

form

oil

potency

2920 mg/kg LD50, oral (Rat)

kinematic viscosity

11.8 cSt(20 °C)

mp

-35 °C

transition temp

flash point 126 °C

density

0.95 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C(C)\C=C\C1=C(CCCC1(C)C)C

InChI

1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h7-8H,5-6,9H2,1-4H3/b8-7+

InChI key

PSQYTAPXSHCGMF-BQYQJAHWSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.944 - 0.952
Identity (IR): passes test

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service