8.15023
Cumene-D12
deuteration degree min. 99% for NMR spectroscopy MagniSolv™
Synonym(s):
Cumene-D12
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
vapor pressure
5.3 hPa ( 20 °C)
form
liquid
autoignition temp.
420 °C (DIN 51794)
potency
1400 mg/kg LD50, oral (Rat)
10578 mg/kg LD50, skin (Rabbit)
expl. lim.
0.8-6.0 % (v/v)
bp
153 °C/1013 hPa
mp
-96 °C
transition temp
flash point 31 °C
density
0.86 g/cm3 at 20 °C
storage temp.
2-30°C
General description
A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.
Analysis Note
Deuteration degree: ≥ 99 %
Legal Information
MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
Flash Point(C)
31 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service