Skip to Content
Merck
All Photos(2)

Key Documents

5.37983

Sigma-Aldrich

Pantothenate Kinase Inhibitor

Synonym(s):

Pantothenate Kinase Inhibitor, 3-(2,4-Dimethylpyrido-[2ʹ,3ʹ:3,4]pyrazolo[1,5-a]pyrimidin-3-yl)-N-(3-(methylthio)phenyl)propanamide, PanK Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N5O
Molecular Weight:
359.42
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

General description

A cell-permeable pyridopyrazolopyrimidine based compound that acts as a potent and reversible inhibitor of pantothenate kinase (IC50 = 70, 92 and 25 nM for PanK1β, PanK2 and PanK3, respectively; radiochemical assay, 2.5 mM [ATP]). Binds to ATP-PanK complex (apparent binding constant ~300 nM for ATP-PanK3) and exhibits mixed-type inhibition with respect to ATP and pantothenate. Shown to dose-dependently decrease [3H]pantothenate incorporation into coenzyme A (apparent IC50 = 900 nM in C3A cells) with negligible off-target toxicity (IC50 >56 µM in Hep-G2 cells).
A cell-permeable pyridopyrazolopyrimidine based compound that acts as a potent and reversible inhibitor of pantothenate kinase (IC50 = 70, 92 and 25 nM for PanK1β, PanK2 and PanK3, respectively; radiochemical assay, 2.5 mM [ATP]). Binds to ATP-PanK complex (apparent binding constant ~300 nM for ATP-PanK3) and exhibits mixed-type inhibition with respect to ATP and pantothenate. Shown to dose-dependently decrease [3H]pantothenate incorporation into coenzyme A (apparent IC50 = 900 nM in C3A cells) with negligible off-target toxicity (IC50 >56 µM in Hep-G2 cells).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
pantothenate kinase
Reversible: yes
Target IC50: 70, 92 and 25 nM for PanK1&beta

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sharma, L.K., et al. 2015. J. Med. Chem.58, 1563.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service