Skip to Content
Merck
All Photos(1)

Key Documents

5.05980

Sigma-Aldrich

Autophagy Inhibitor IV, Cpd18

Synonym(s):

Autophagy Inhibitor IV, Cpd18, 3-methyl-6-(3-methylpiperidin-1-yl)-3H-purine, 3-MA analog, Cpd 18

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17N5
Molecular Weight:
231.30
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

0.67 mM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable 3-methyladenine (3-MA) derivative that acts as an autophagy inhibitor via the inhibition of class III PI3K, and without touching class I PI3K. Shown to block 80% of starvation - induced autophagy at 1 mM (IC50 = 0.67 mM, compared with 3-MA at 6 mM). Displays much better solubility than 3-MA (5 times more soluble in water and 30 times more soluble in DMSO). Also found to reduce cell viability by 55% at 1 mM at 12 hours compared with 72% reduction for 3-MA at 10 mM.
A cell-permeable 3-methyladenine (3-MA) derivative that acts as an autophagy inhibitor via the inhibition of class III PI3K, and without touching class I PI3K. Shown to block 80% of starvation-induced autophagy at 1 mM (IC50 = 0.67 mM, compared with 3-MA at 6 mM). Displays much better solubility than 3-MA (5 times more soluble in water and 30 times more soluble in DMSO). Also found to reduce cell viability by 55% at 1 mM at 12 hours compared with 72% reduction for 3-MA at 10 mM.

Please note that the molecular weight for this compound is lot-specific due to variable water content.

Biochem/physiol Actions

Primary Target
PI3K
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wu, Y., et al, 2013. Autophagy9, 595.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service