Skip to Content
Merck
All Photos(1)

Key Documents

1.44209

Millipore

Tripod

for use with RCS™ High Flow, RCS™ High Flow Touch, RCS™ Plus Ex

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409

material

metal stand

Agency

non-compendial

feature

adjustable

packaging

pkg of 1 kit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

storage temp.

room temp

General description

Tripod is an additional accessory for use with RCS® microbial air sampler. The RCS®portable air sampler is used for fast and easy monitoring of ambient air and compressed gases in controlled environments. The microbial air sampler is based on impaction by centrifugal force. The Tripod is used for microbial air sampling in heights up to three meters.

Application

Tripod is an additional accessory with the RCS® microbial air sampler used for sampling in heights up to three meters. These air samplers find application in microbial monitoring of ambient air in pharmaceutical, medical and food industry.

Compatibility

The Tripod is compatible for use with RCS® Air Samplers of the High Flow series (RCS® High Flow Touch, RCS® High Flow) and RCS®Isolator.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service