1.17171
Petridish Clamps
size 4 mm, For MAS-100 VF®, 3 Nos
About This Item
Recommended Products
material
aluminum lid
Agency
non-compendial
description
connector
feature
autoclavable
packaging
pkg of 1 kit
size
4 mm
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 VF®
storage temp.
no temp limit
General description
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service