Skip to Content
Merck
All Photos(1)

Key Documents

1.07531

Supelco

1-(2-Pyridylazo)-2-naphthol

(PAN) metal indicator, Reag. Ph Eur

Synonym(s):

1-(2-Pyridylazo)-2-naphthol (PAN), PAN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N3O
CAS Number:
Molecular Weight:
249.27
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-637-9
NACRES:
NB.21

Product Name

1-(2-Pyridylazo)-2-naphthol (PAN), metal indicator Reag. Ph Eur

Agency

reag. Ph. Eur.

Quality Level

form

solid

color

brick red to orange-brown
clear to almost clear red-orange to brown-orange, 1.0g/L;ethanol

mp

136-141 °C

bulk density

190 kg/m3

λmax

461-465 nm in ethanol

storage temp.

2-30°C

SMILES string

N(\N=C2\c3c(cccc3)C=CC\2=O)c1ncccc1

InChI

1S/C15H11N3O/c19-13-9-8-11-5-1-2-6-12(11)15(13)18-17-14-7-3-4-10-16-14/h1-10H,(H,16,17)/b18-15-

InChI key

RAXUMGMWXZYADR-SDXDJHTJSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Derivative spectrophotometric determination of iridium after preconcentration of its 1-(2-pyridylazo)-2-naphthol complex on microcrystalline naphthalene.: This study explores a spectrophotometric method for determining iridium using 1-(2-Pyridylazo)-2-naphthol (PAN) as a complexing agent, demonstrating an effective approach for trace metal analysis in academic and research applications, highlighting its relevance to environmental monitoring and analytical chemistry (Taher MA et al., 1997).

  • Simultaneous determination of gallium(III) and indium(III) by derivative spectrophotometry.: This research presents a derivative spectrophotometric technique for the concurrent determination of gallium and indium, employing PAN for chelation, underscoring its utility in the precise analysis of these metals in various sample matrices, which is crucial for chemists working in analytical fields (Singh VK et al., 2001).

  • Second and first-derivative spectrophotometry for efficient simultaneous and individual determination of palladium and cobalt using 1-(2-pyridylazo)-2-naphthol in sodium dodecylsulfate micellar media.: This paper introduces a novel approach for the simultaneous spectrophotometric determination of palladium and cobalt using PAN, offering significant advantages in terms of sensitivity and selectivity, essential for advancing analytical methodologies in chemistry (Eskandari H et al., 2003).

  • Zero-crossing second derivative spectrophotometry for simultaneous determination of palladium and nickel using 1-(2-pyridylazo)-2-naphthol in mixed micellar solutions.: This publication details a method for analyzing palladium and nickel using PAN, employing advanced spectrophotometric techniques to enhance detection capabilities, reflecting critical developments in analytical techniques relevant to chemists in academic settings (Eskandari H, 2004).

  • Preconcentration of 1-(2-pyridylazo)-2-naphthol-iron(III)-capriquat on a membrane filter, and third-derivative spectrophotometric determination of iron(III).: The study focuses on a preconcentration technique for iron analysis using PAN, demonstrating the application of derivative spectrophotometry in enhancing analytical precision, an essential aspect for researchers focused on trace metal analysis in environmental and industrial samples (Mori I et al., 1994).

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Appearance: brick-red to orange-brown fine powder,eventually cohesive
Appearance of solution (1.0 g/l; ethanol): Clear to almost clear, red-orange to brown-orange
Melting point: 136 - 141 °C
Absorption maximum λmax. (Ethanol): 461 - 465 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; Ethanol): 670 - 720
Suitability as indicator (for metal titration): passes test
Sensitivity test (Reag. Ph Eur): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service