All Photos(1)
About This Item
Linear Formula:
(CD3)3COCD3
Molecular Weight:
100.22
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
99 atom % D
Quality Level
Assay
99% (CP)
bp
55-56 °C
density
0.840 g/mL at 25 °C
mass shift
M+12
storage temp.
−20°C
SMILES string
[2H]C([2H])([2H])OC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
InChI
1S/C5H12O/c1-5(2,3)6-4/h1-4H3/i1D3,2D3,3D3,4D3
InChI key
BZLVMXJERCGZMT-MGKWXGLJSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-18.4 °F - closed cup
Flash Point(C)
-28 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service