Merck
  • Antigen-induced regulation of T-cell motility, interaction with antigen-presenting cells and activation through endogenous thrombospondin-1 and its receptors.
Antigen-induced regulation of T-cell motility, interaction with antigen-presenting cells and activation through endogenous thrombospondin-1 and its receptors.
Immunology (2014-11-14)
Sten-Erik Bergström, Mehmet Uzunel, Toomas Talme, Eva Bergdahl, Karl-Gösta Sundqvist
ZUSAMMENFASSUNG
Antigen recognition reduces T-cell motility, and induces prolonged contact with antigen-presenting cells and activation through mechanisms that remain unclear. Here we show that the T-cell receptor (TCR) and CD28 regulate T-cell motility, contact with antigen-presenting cells and activation through endogenous thrombospondin-1 (TSP-1) and its receptors low-density lipoprotein receptor-related protein 1 (LRP1), calreticulin and CD47. Antigen stimulation induced a prominent up-regulation of TSP-1 expression, and transiently increased and subsequently decreased LRP1 expression whereas calreticulin was unaffected. This antigen-induced TSP-1/LRP1 response down-regulated a motogenic mechanism directed by LRP1-mediated processing of TSP-1 in cis within the same plasma membrane while promoting contact with antigen-presenting cells and activation through cis interaction of the C-terminal domain of TSP-1 with CD47 in response to N-terminal TSP-1 triggering by calreticulin. The antigen-induced TSP-1/LRP1 response maintained a reduced but significant motility level in activated cells. Blocking CD28 co-stimulation abrogated LRP1 and TSP-1 expression and motility. TCR/CD3 ligation alone enhanced TSP-1 expression whereas CD28 ligation alone enhanced LRP1 expression. Silencing of TSP-1 inhibited T-cell conjugation to antigen-presenting cells and T helper type 1 (Th1) and Th2 cytokine responses. The Th1 response enhanced motility and increased TSP-1 expression through interleukin-2, whereas the Th2 response weakened motility and reduced LRP1 expression through interleukin-4. Ligation of the TCR and CD28 therefore elicits a TSP-1/LRP1 response that stimulates prolonged contact with antigen-presenting cells and, although down-regulating motility, maintains a significant motility level to allow serial contacts and activation. Th1 and Th2 cytokine responses differentially regulate T-cell expression of TSP-1 and LRP1 and motility.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, suitable for HPLC
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Wasser, Deionized
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Wasser, for molecular biology, sterile filtered
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Wasser, BioPerformance Certified
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Wasser, ACS reagent
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Wasser, PCR Reagent
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Wasser, endotoxin, free
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
4-Brommethyl-7-methoxycumarin, 97%
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Supelco
Wasser, ACS reagent, for ultratrace analysis
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Supelco
Wasser, suitable for ion chromatography
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Supelco
Wasser, for TOC analysis
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Wasser, tested according to Ph. Eur.
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Supelco
Dichte Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Anti-Pigmentepithel abgeleiteter Faktor-Antikörper, Klon 10F12.2, clone 10F12.2, Chemicon®, from mouse
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Supelco
Wasser, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge