Merck
Effects of histamine H4 receptor ligands in a mouse model of gastric ulceration.
Pharmacology (2012-04-28)
Maristella Adami, Cristina Pozzoli, Alessandro Menozzi, Simone Bertini, Benedetta Passeri, Anna Maria Cantoni, Rogier Smits, Iwan de Esch, Rob Leurs, Gabriella Coruzzi
ZUSAMMENFASSUNG
In the present study we examined whether histamine H(4) receptors (H(4)Rs) have a role in gastric ulcerogenesis using a mouse model of gastric damage. The H(4)R antagonist JNJ7777120 and the H(4)R agonists VUF8430 and VUF10460 were investigated in fasted CD-1 mice against the ulcerogenic effect induced by co-administration of indomethacin(IND, 30 mg/kg s.c.) and bethanechol (BET, 5 mg/kg i.p.). Both macroscopic and histologic lesions were examined. Strain-related differences were investigated by testing JNJ7777120 also in NMRI, BALB/c and C57BL/6J mice. Neither JNJ7777120 nor the H(4)R agonists displayed effects in the normal stomach at any dose tested (10 and 30 mg/kg s.c.). As expected, IND+BET provoked several lesions in the fundic mucosa, which were significantly reduced by JNJ7777120 (10 and 30 mg/kg s.c.). The gastroprotective effect of JNJ7777120 (10 and 30 mg/kg s.c.) was observed in CD-1, NMRI and BALB/c, but not in C57BL/6J, mice. In CD-1 mice, the H(4)R agonists VUF8430 and VUF10460 (both at 10 and 30 mg/kg s.c.) did not modify the damage induced by IND+BET, however VUF8430 (10 mg/kg s.c.) prevented the gastroprotection induced by JNJ7777120 (10 mg/kg s.c.). Data obtained with selective ligands suggest that the H(4)R may have a role in mouse gastric ulcerogenesis. If confirmed in humans, these data would emphasize the potential advantage of H(4)R blockers as gastrosparing anti-inflammatory drugs. The lack of effects of JNJ7777120 in C57BL/6J mice has to be carefully considered in the pharmacological characterization of H(4)R functions and/or new selective ligands.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carbamyl-β-Methylcholinchlorid, ≥99% (TLC), crystalline