Merck
509485
SGE gasdichte Spritzen, Luer-Lock
500F-LL-GT, volume 500 μL, needle size (not included)
Synonym(e):
SPRITZE SGE 500μL, LUER LOCK
About This Item
UNSPSC-Code:
41122001
Materialien
glass syringe
Kanülengröße
(not included)
Leistungsmerkmale
Fixed Luer Lock
Verpackung
pkg of 1 ea
Hersteller/Markenname
SGE 007230
Volumen
500 μL
Allgemeine Beschreibung
SGE gas tight syringe without a needle, Model: 500F-LL-GT (volume 0.5 mL)
  • Made of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
  • Autoclavable syringes but the plunger should be removed.
  • Features an interference fit between the plunger tip and the barrel which enhances the gas sealing properties of the syringe.
  • Maximum inertness for liquid or gas samples.
  • Suitable for manual dispenser/diluter applications.
  • High accuracy of dispensed volume.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)