Ansicht ändern
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
UNSPSC Code:
41115719
NACRES:
SB.54
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenpackaging
pkg of 5 ea
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
91.5 mm × 3.0 mm × 1.8 mm
General description
- Länge 91.5 mm × AD 3 mm × ID 1.8 mm
- Ersetzt die Herst.-Artikel-Nr.: 5080-3782
The liner is the second most important part of the inlet. It is the centrepiece of the inlet system, basically a glass tube between the septum and the column, where the sample is evaporated and brought into the gas phase. There are three most important parameters one should keep in mind while choosing a liner, firstly liner volume, considering a typical liner has a volume of 900μl, secondly liner treatment, one should always use a deactivated liner, so that the glass does not react with the analytes introduced in the liner, thirdly liner design, different designs are available like straight, tapered, baffled and without glass wool and one should be careful enough while choosing the design such that it is compatible with the desired injection mode.
Application
Inlet liner maybe used for the determination and evaluation of volatile components in food products. The vegetable oil maybe directly placed into the GC inlet liner packed with glass wool and then inserted into the heated inlet of the GC for analysis.
1 of 1
Dieser Artikel | ||
|---|---|---|
| packaging pkg of 5 ea | packaging pkg of 25 ea | packaging pkg of 5 ea |
| technique(s) gas chromatography (GC): suitable | technique(s) gas chromatography (GC): suitable | technique(s) - |
| L × O.D. × I.D. 91.5 mm × 3.0 mm × 1.8 mm | L × O.D. × I.D. 91.5 mm × 3.0 mm × 1.8 mm | L × O.D. × I.D. 112 mm × 6.0 mm × 3.0 mm |
Still not finding the right product?
Explore all of our products under Inlet-Liner für gepackte GC (ungepackt)
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?