Merck
Z763152
Prism centrifuge adapters
for 0.2 mL tubes, pack of 8 ea
Synonym(e):
lab equipment accessory
About This Item
UNSPSC-Code:
41103911
NACRES:
NB.22
Leistungsmerkmale
Holds 0.2 mL tubes
Verpackung
pack of 8 ea
Hersteller/Markenname
Labnet C1222-8
Methode(n)
centrifugation: suitable
Kompatibilität
for use with C1801
for use with Labnet Prism (TM) Mini Centrifuge
Rechtliche Hinweise
Prism is a trademark of Labnet Intl., Inc.