Z756911
Universal animal restrainer
small, pack of 1 ea
Synonym(e):
animal container
Empfohlene Produkte
Materialien
clear acrylic
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art H46400-0001
Allgemeine Beschreibung
Holds animal: mice
A variety of small animals and rodents can be restrained with this unit simply by moving the head gate to one of 4 different positions. To accommodate larger rodents, simply pull out the inner floor, increasing the available room. Stable base legs are provided by milled, chrome-plated brass thumb screws
Minimum restraining: 5.1 x 2.9 cm (2 x 1 1/8 in.)
Maximum restraining: 10.2 x 2.9 cm (4 x 1 1/8 in.)
A variety of small animals and rodents can be restrained with this unit simply by moving the head gate to one of 4 different positions. To accommodate larger rodents, simply pull out the inner floor, increasing the available room. Stable base legs are provided by milled, chrome-plated brass thumb screws
- Humanely restrain small animals
- Comes with vented head gate and tail gate that are easily removed
- Occlusion cuff held close to the animal′s body with the tailgate
Minimum restraining: 5.1 x 2.9 cm (2 x 1 1/8 in.)
Maximum restraining: 10.2 x 2.9 cm (4 x 1 1/8 in.)