Merck
Z719668
Sorenson low binding standard tips
maximum volume 1000 μL, pack of 1000 ea (Bag), non-sterile
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
Maximales Volumen:
1000 μL
Sterilität:
non-sterile
Leistungsmerkmale:
barrier: no
Verpackung:
pack of 1000 ea (Bag)
case of 10000 ea (10 bags of 100 tips)
Hersteller/Markenname:
Sorenson 10190T
Materialien
colorless
polypropylene
Sterilität
non-sterile
Leistungsmerkmale
barrier: no
Verpackung
pack of 1000 ea (Bag)
case of 10000 ea (10 bags of 100 tips)
Hersteller/Markenname
Sorenson 10190T
Maximales Volumen
1000 μL
Allgemeine Beschreibung
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
Rechtliche Hinweise
Sorenson is a trademark of Sorenson BioScience, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)