Merck
Z717401
Nichiryo pipette tips
volume range 1-10 mL, universal, bulk pack
Synonym(e):
Disposable Pipette Tips, Precision Pipette Tips
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
Materialien
clear tip
colorless
Sterilität
non-sterile
Leistungsmerkmale
DNase free
RNase free
barrier: no
Verpackung
pack of 200 ea (Bag)
Hersteller/Markenname
Nichiryo BMT-Z
Verfügbarkeit
not available in (France)
Maximales Volumen
10 mL
Volumenbereich
1-10 mL
Leistungsmerkmale und Vorteile
  • RNase and DNase free
  • Fits most pipettes
  • Premium packaging featuring stand-up and puncture-resistant bags
  • Racked tips are offered in both hinged and lift-off tops
Sonstige Hinweise
Browse all pipette tips